Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Lieferant: Honeywell Chemicals
65758-25L
HONC65758-25LEA
1446.53
EUR
HONC65758-25L
Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Ethanol
Absolute alcohol, sales not in Germany, for license holders only
Formel:
C₂H₅OH Siedepunkt: 78…79 °C (1,013 hPa) Schmelzpunkt: –115 °C Dichte: 0,790…0.791 g/cm3 (20 °C) Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...