n-Hexan ACS, Reag. Ph. Eur., Burdick & Jackson™
Lieferant: Honeywell Chemicals
65801-200L
HONC65801-200LEA
7988.38
EUR
HONC65801-200L
n-Hexan ACS, Reag. Ph. Eur., Burdick & Jackson™
n-Hexan
Formel:
H₃C(CH₂)₄CH₃ Siedepunkt: 66…69 °C Dichte: 0,660…0.668 g/cm3 Flammpunkt: –26 °C |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 UN: 1208 ADR: 3,II |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 99 % |
Boiling range | 68 - 69 °C |
Boiling range 67-69 °C | Min. 95 % |
Density (D 20/4) | 0.658 - 0.662 |
Density (D 20/20) | 0.659 - 0.663 |
Refractive index (n 20/D) | 1.3745 - 1.3755 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.01 % |
Free acid (as CH₃COOH) | Max. 0.002 % |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.00001 % |
Tin (Sn) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
Aromatic substances (as C₆H₆) | Max. 0.01 % |
S-compounds (as S) | Max. 0.005 % |
Thiophene | Max. 0.0001 % |
Reaction against H₂SO₄ | complying |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...