D(+)-Saccharose, Extra rein erfüllt analytische Spezifikationen von Reagent Ph. Eur., BP, NF, Burdick & Jackson™
Lieferant: Honeywell Chemicals
65893-10KG
HONC65893-10KGEA
354.21
EUR
HONC65893-10KG
D(+)-Saccharose, Extra rein erfüllt analytische Spezifikationen von Reagent Ph. Eur., BP, NF, Burdick & Jackson™
D(+)-Saccharose
Formel:
C₁₂H₂₂O₁₁ Schmelzpunkt: 185…186 °C Dichte: 1,580 g/cm3 (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 EINECS: 200-334-9 |
|
Spezifikation Testergebnisse
Identity | complying |
Specific rotation (a)20/D;C=26 in H₂O | 66.3 - 67.0 ° |
Conductivity 20 °C | Max. 35 µS/cm |
Loss on drying (105°C, 3h) | Max. 0.1 % |
Residue on ignition (SO₄) | Max. 0.05 % |
Acidity or alkalinity | complying |
Calcium (Ca) | complying |
Lead (Pb) | Max. 0.5 ppm |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.0035 % |
Sulfate (SO₄) | Max. 0.006 % |
Sulfite (as SO₂) | Max. 0.001 % |
Dextrines | complying |
Invert sugar | complying |
Reducing sugars | complying |
Colour index | Max. 45 |
Appearance of the solution | complying |
Residual solvents | complying |
Endotoxin limit (LAL) | Max. 250 EU/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...