Natriumdihydrogenphosphat Monohydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Natriumphosphat monobasisch Monohydrat

71504-250G 71504-1KG 71504-25KG
71504-250GEA 40.12 EUR
71504-250G 71504-1KG HONC71504-25KG
Natriumdihydrogenphosphat Monohydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Molecular Weight: 137.99 g/mol
Schmelzpunkt: 650 °C (wasserfrei)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
pH (5 %, 25 °C) 4.1 - 4.5
Colour of the solution complying
Turbidity of solution complying
Water insoluble matter Max. 0.01 %
Arsenic (As) Max. 0.5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm
Total nitrogen (as N) Max. 10 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR