Zinkchlorid, Spezialqualität

Lieferant: Honeywell Chemicals
Gefahr

72186-25KG
HONC72186-25KGEA 2193.91 EUR
HONC72186-25KG
Zinkchlorid, Spezialqualität
Zinkchlorid
Formel: ZnCl₂
Dichte: 2,93 g/cm3
MDL: MFCD00011295
CAS-Nummer: 7646-85-7
UN: 2331
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Assay 98.0 - 100.5 %
Oxide chloride (as ZnO) complying
pH (10 %, 20°C) 4.6 - 5.3
Ammonium (NH₄) Max. 0.03 %
Arsenic (As) Max. 0.0002 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.02 %
Magnesium (Mg) Max. 0.01 %
Sodium (Na) Max. 0.01 %
Lead (Pb) Max. 0.005 %
Total N Max. 0.005 %
Sulfate (SO₄) Max. 200 ppm
Al, Ca, Fe, Mg and heavy metals complying
Alkali and alkaline earth Max. 1.0 %
Residual solvents complying
Turbidity Max. 3.5 NTU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR