Perchlorsäure ≥60% (durch titrimetrische Analyse), Puriss. p.a. ACS analytical reagent, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Überchlorsäure
Packed in coated, shock-and leak-protected glass bottle
77234-1L
77234-500ML
77234-1LEA
202.49
EUR
77234-1L
HONC77234-500ML
Perchlorsäure ≥60% (durch titrimetrische Analyse), Puriss. p.a. ACS analytical reagent, Fluka™
Perchlorsäure
Packed in coated, shock- and leak-protected glass bottle
Formel:
HClO₄ Siedepunkt: 203 °C(1,013 hPa) Schmelzpunkt: –18 °C Dichte: 1,67 g/cm³ Flammpunkt: 113 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00011325 CAS-Nummer: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1,I |
|
Spezifikation Testergebnisse
Assay | 60.0 - 62.0 % |
Density (D 20/4) | 1.536 - 1.558 |
Residue on ignition | Max. 30 ppm |
Silver (Ag) | Max. 0.1 ppm |
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.05 ppm |
Barium (Ba) | Max. 0.02 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.05 ppm |
Copper (Cu) | Max. 0.1 ppm |
Iron (Fe) | Max. 1 ppm |
Potassium (K) | Max. 1 ppm |
Lithium (Li) | Max. 0.02 ppm |
Magnesium (Mg) | Max. 0.5 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.05 ppm |
Nickel (Ni) | Max. 0.1 ppm |
Lead (Pb) | Max. 0.05 ppm |
Strontium (Sr) | Max. 0.02 ppm |
Thallium (Tl) | Max. 0.05 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Heavy metals (as Pb) | Max. 0.0001 % |
Phosphate, silicate (as SiO₂) | Max. 5 ppm |
Chloride (Cl) | Max. 3000 ppb |
Sulfate (SO₄) | Max. 10 ppm |
Free chlorine (Cl) | Max. 0.0001 % |
Chlorate (ClO₃) | Max. 10 ppm |
Total N | Max. 10 ppm |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...