Capping Reagens, BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

BC644-1200KG BC644-2.5L
HONCBC644-1200KGEA 135074.69 EUR
HONCBC644-1200KG HONCBC644-2.5L
Capping Reagens, BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Capping Reagens

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Dichte: 0,881 g/cm3 (20 °C)
Flammpunkt: 11 °C
Lagertemperatur: Raumtemperatur
EINECS: 200-835-2
UN: 2924
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR