Trichloressigsäure 3% (w/v) in Dichlormethan, BioSyn™ Detritylierungsreagenz, zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: TCA

BR605-4X2.5L
HONCBR605-4X2.5LEA 569.02 EUR
HONCBR605-4X2.5L
Trichloressigsäure 3% (w/v) in Dichlormethan, BioSyn™ Detritylierungsreagenz, zur DNA/RNA Synthese, Burdick & Jackson™
Trichloressigsäure

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


UNSPSC Code: 12350000

Formel: Cl₃CCOOH
Molecular Weight: 163,39 g/mol
Siedepunkt: 40 °C
Schmelzpunkt: -95 °C
Dichte: 1,33 g/cm³ (20 °C)
Flammpunkt: Does not flash, Closed cup (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00004177
CAS-Nummer: 76-03-9
EINECS: 200-838-9
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR