Capping Reagens 90/10 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

BR651-4X2.5L
HONCBR651-4X2.5LEA 3023.05 EUR
HONCBR651-4X2.5L
Capping Reagens 90/10 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Capping Reagens

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


UNSPSC Code: 12350000

Siedepunkt: 66 °C (Hauptbestandteil)
Dichte: 0,888 g/cm³ (20 °C) (Hauptbestandteil)
Flammpunkt: –15 °C
Lagertemperatur: Raumtemperatur
UN: 2924
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR