Oxidationsreagenz 0.02 M I₂ in 78/20/2 THF/Pyridin/Wasser (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

BR663-4X2.5L
HONCBR663-4X2.5LEA 1433.96 EUR
HONCBR663-4X2.5L
Oxidationsreagenz 0.02 M I₂ in 78/20/2 THF/Pyridin/Wasser (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Iod

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


DNA reagents specifically processed and purified for lab scale oligonucleotide synthesis. Suitable for ABI 380 and 390 series and 3948 DNA/RNA synthesizers. Mixture passed through 0.2 µm filter prior to packaging to minimize particulate contamination.

Formel: I₂
Molecular Weight: 253.81 g/mol
Siedepunkt: 66 °C
Schmelzpunkt: –108.5 °C
Dichte: 0.888 g/cm3
Flammpunkt: –15 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
UN: 1993
ADR: 3 II
Merck Index: 13,05036

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR