Oxidationsreagenz 0.02 M I₂ in 70/20/10 THF/Pyridin/Wasser (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

BR666-4X2.5L
HONCBR666-4X2.5LEA 1296.8 EUR
HONCBR666-4X2.5L
Oxidationsreagenz 0.02 M I₂ in 70/20/10 THF/Pyridin/Wasser (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Iod

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


UNSPSC Code: 12350000

Formel: I₂
Siedepunkt: 66 °C
Schmelzpunkt: –108,5 °C
Dichte: 0,935 g/cm3 (20 °C)
Flammpunkt: –15 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR