5-(Ethylthio)-1H-tetrazol (ETT) 0.5M in Acetonitril, BioSyn™ Aktivatorreagenz, zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: 5-Ethyltio-1H-Tetrazole

BR725-4X2.5GL
HONCBR725-4X2.5GLEA 14004.41 EUR
HONCBR725-4X2.5GL
5-(Ethylthio)-1H-tetrazol (ETT) 0.5M in Acetonitril, BioSyn™ Aktivatorreagenz, zur DNA/RNA Synthese, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazol (ETT)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Formel: C₃H₆N₄S
Molecular Weight: 130,17 g/mol
Siedepunkt: 82 °C (main component)
Schmelzpunkt: -46 °C (main component)
Dichte: 0,809 g/cm³ (20 °C)
Flammpunkt: 13 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068733
CAS-Nummer: 89797-68-2
UN: 1648
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR