Gold ICP Standard, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1006-100ML
HONCE1006-100MLEA
0
EUR
HONCE1006-100ML
Gold ICP Standard, (CRMs), Fluka™
Standards
Kationischer Standards
Gold-Standards
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).