Erbium ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1019-100ML
E1146-100ML
HONCE1019-100MLEA
111.58
EUR
HONCE1019-100ML
HONCE1146-100ML
Erbium ICP standards, (CRMs), Fluka™
Standards
Kationischer Standards
Erbium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).