Gadolinium ICP Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1023-100ML
HONCE1023-100MLEA
112.26
EUR
HONCE1023-100ML
Gadolinium ICP Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Gadolinium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).