Platinum ICP Standard, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1046-100ML
HONCE1046-100MLEA 301.42 EUR
HONCE1046-100ML
Platinum ICP Standard, (CRMs), Fluka™
Standards Kationischer Standards Platin-Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).