Samarium ICP Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1056-100ML
HONCE1056-100MLEA 169.25 EUR
HONCE1056-100ML
Samarium ICP Standards, (CRMs), Fluka™
Standards Kationischer Standards Samarium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).