Boron ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1134-100ML
HONCE1134-100MLEA
138.13
EUR
HONCE1134-100ML
Boron ICP standards, (CRMs), Fluka™
Standards
Kationischer Standards
Bor-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).