Cadmium ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1139-100ML
HONCE1139-100MLEA
135.64
EUR
HONCE1139-100ML
Cadmium ICP standards, (CRMs), Fluka™
Standards
Kationischer Standards
Cadmium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).