Magnesium ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1034-100ML
E1161-100ML
HONCE1034-100MLEA
77.57
EUR
HONCE1034-100ML
HONCE1161-100ML
Magnesium ICP standards, (CRMs), Fluka™
Standards
Kationischer Standards
Magnesium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).