Lead ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1170-100ML
HONCE1170-100MLEA 134.11 EUR
HONCE1170-100ML
Lead ICP standards, (CRMs), Fluka™
Standards Kationischer Standards Blei-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).