Gallium ICP & ICP-MS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1319-100ML E1387-100ML
HONCE1319-100MLEA 96.66 EUR
HONCE1319-100ML HONCE1387-100ML
Gallium ICP & ICP-MS Standards, (CRMs), Fluka™
Standards Kationischer Standards Gallium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).