Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1326-100ML
HONCE1326-100MLEA 171.45 EUR
HONCE1326-100ML
Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Standards Kationischer Standards Iridium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).