Magnesium ICP and ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1331-100ML E1399-100ML
HONCE1331-100MLEA 0 EUR
HONCE1331-100ML HONCE1399-100ML
Magnesium ICP and ICP-MS standards, (CRMs), Fluka™
Standards Kationischer Standards Magnesium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).