Tantalum ICP & ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1355-100ML E1423-100ML
HONCE1355-100MLEA 110.48 EUR
HONCE1355-100ML HONCE1423-100ML
Tantalum ICP & ICP-MS standards, (CRMs), Fluka™
Standards Kationischer Standards Tantal-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).