Boron ICP & ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1304-100ML E1373-100ML
HONCE1304-100MLEA 96.66 EUR
HONCE1304-100ML HONCE1373-100ML
Boron ICP & ICP-MS standards, (CRMs), Fluka™
Standards Kationischer Standards Bor-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).