Boron ICP & ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1304-100ML
E1373-100ML
HONCE1304-100MLEA
96.66
EUR
HONCE1304-100ML
HONCE1373-100ML
Boron ICP & ICP-MS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Bor-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).