Barium ICP and ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1374-100ML
HONCE1374-100MLEA 94.03 EUR
HONCE1374-100ML
Barium ICP and ICP-MS standards, (CRMs), Fluka™
Standards Kationischer Standards Barium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).