Indium ICP & ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1325-100ML
E1393-100ML
HONCE1325-100MLEA
110.48
EUR
HONCE1325-100ML
HONCE1393-100ML
Indium ICP & ICP-MS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Indium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).