Zinc ICP and ICP-MS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1366-100ML
E1435-100ML
HONCE1366-100MLEA
0
EUR
HONCE1366-100ML
HONCE1435-100ML
Zinc ICP and ICP-MS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Zink-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).