Initial calibration verification standard for ICP-MS
Lieferant: Honeywell Chemicals
E2003-100ML
HONCE2003-100MLEA
930.83
EUR
HONCE2003-100ML
Initial calibration verification standard for ICP-MS
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
1000 mg/L each of Ca, Fe, K, Mg, Na, Sr, 10 mg/L each of Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Tl, V, Zn, Th, U in HNO₃ 5%