Interference check solution A for ICP-MS systems
Lieferant: Honeywell Chemicals
E2004-100ML
HONCE2004-100MLEA
635.92
EUR
HONCE2004-100ML
Interference check solution A for ICP-MS systems
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
20 g/L Cl-, 3000 mg/L Ca, 2500 mg/L each of Fe, Na; 2000 mg/L C, 1000 mg/L each of Al, Mg, P, K, S, 20 mg/L each of Mo, Ti in HNO₃ 5%