Internal standard mix for ICP-MS systems
Lieferant: Honeywell Chemicals
E2006-100ML
HONCE2006-100MLEA 447.99 EUR
HONCE2006-100ML
Internal standard mix for ICP-MS systems
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Bi, Ge, In, 6Li, Lu, Rh, Sc, Tb in HNO₃ 10%