ICP-OES Wavelength calibration Concentrate
Lieferant: Honeywell Chemicals
E2015-100ML
HONCE2015-100MLEA 710.59 EUR
HONCE2015-100ML
ICP-OES Wavelength calibration Concentrate
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
500mg/L K, 50 mg/L each of Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn in HNO₃ 5%