Calibration mix 1 for AA and ICP-OES
Lieferant: Honeywell Chemicals
E2020-100ML
HONCE2020-100MLEA
382.04
EUR
HONCE2020-100ML
Calibration mix 1 for AA and ICP-OES
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Mo, Sb, Sn, Ti in HNO₃ 2%, HF 0.5%