Calibration mix 2 for AA and ICP-OES
Lieferant: Honeywell Chemicals
E2021-100ML
HONCE2021-100MLEA
412.04
EUR
HONCE2021-100ML
Calibration mix 2 for AA and ICP-OES
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Ni, Pb, Se, Th, Tl, U, V, Zn in HNO₃ 5%