Combined Seven Anion Standard II for IC
Lieferant: Honeywell Chemicals
E2030-100ML
HONCE2030-100MLEA 308.35 EUR
HONCE2030-100ML
Combined Seven Anion Standard II for IC
Standards Anionische Standards Anionische Multi-Element-Standards
Certified Reference Material (CRM).
200 mg/L PO43-, 100 mg/L each of Cl-, NO2-, Br-, NO3-, SO42-, 20 mg/L F- in H₂O