Combined Five Anion Standard for IC
Lieferant: Honeywell Chemicals
E2031-100ML
HONCE2031-100MLEA 0 EUR
HONCE2031-100ML
Combined Five Anion Standard for IC
Standards Anionische Standards Anionische Multi-Element-Standards
Certified Reference Material (CRM).
150 mg/L each of PO43-, SO42-, 100 mg/L NO3-, 30 mg/L Cl-, 20 mg/L F- in H₂O
Bestellen
TECHNISCHE DATEN
Environmentally Preferable