Mixed calibration standard 2 for ICP
Lieferant: Honeywell Chemicals
E2095-100ML
HONCE2095-100MLEA
752.19
EUR
HONCE2095-100ML
Mixed calibration standard 2 for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
10 g/L Fe, 100mg/L each of Ba, Co, Cu, V in HNO₃ 5%