Mixed calibration standard 5 for ICP
Lieferant: Honeywell Chemicals
E2098-100ML
HONCE2098-100MLEA
391.31
EUR
HONCE2098-100ML
Mixed calibration standard 5 for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
1000 mg/L Mg, 200 mg/L each of Sb, Tl, 100 mg/L B, 50 mg/L Ag in HNO₃ 5%