UV Wavecal solution for ICP
Lieferant: Honeywell Chemicals
E2099-100ML
HONCE2099-100MLEA
220.96
EUR
HONCE2099-100ML
UV Wavecal solution for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of K, P, S, 20 mg/L each of As, La, Li, Mn, Mo, Na, Ni, Sc in HCl 2%