Quality control standard pure XVI for ICP
Lieferant: Honeywell Chemicals
E2101-100ML
HONCE2101-100MLEA
366.15
EUR
HONCE2101-100ML
Quality control standard pure XVI for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in HNO₃ 5%