Quality control standard 7A elements for ICP
Lieferant: Honeywell Chemicals
E2102-100ML
HONCE2102-100MLEA
215.7
EUR
HONCE2102-100ML
Quality control standard 7A elements for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
1000 mg/L K, 500 mg/L Si, 100 mg/L each of Al, B, Ba, Na, 50 mg/L Ag in HNO₃ 5%