Instrument Calibration standard 4 for ICP and HGA elements
Lieferant: Honeywell Chemicals
E2106-100ML
HONCE2106-100MLEA
185.7
EUR
HONCE2106-100ML
Instrument Calibration standard 4 for ICP and HGA elements
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of As, Tl, 50 mg/L each of Cd, Se, 30 mg/L Pb in HNO₃ 5%