EPA 6010 interference check standard for ICP-AES
Lieferant: Honeywell Chemicals
E2112-100ML
HONCE2112-100MLEA
778.46
EUR
HONCE2112-100ML
EPA 6010 interference check standard for ICP-AES
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
6000 mg/L Ca, 5000 mg/L Fe, 3000 mg/L Mg, 1200 mg/L Al, 1000 mg/L Na in HNO₃ 5%