Alternate Interference A for ICP
Lieferant: Honeywell Chemicals
E2115-100ML
HONCE2115-100MLEA
626.92
EUR
HONCE2115-100ML
Alternate Interference A for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
1000 mg/L each of Cr, Cu, Mn, Ni, Ti, V in HNO₃ 5%