Alternate Analytes B for ICP
Lieferant: Honeywell Chemicals
E2117-100ML
HONCE2117-100MLEA 542.03 EUR
HONCE2117-100ML
Alternate Analytes B for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Al, As, B, Mo, Na, Sb, Se, Tl, 10 mg/L each of Ca, Fe, Mg, Si in HNO₃ 5%