Analytes C for ICP
Lieferant: Honeywell Chemicals
E2120-100ML
HONCE2120-100MLEA 602.86 EUR
HONCE2120-100ML
Analytes C for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
2 mg/L each of Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg, Mn, Ni, Pb, Sb, Se, Tl, V, Zn in HNO₃ 2%