Multi-element solution 5 for ICP
Lieferant: Honeywell Chemicals
E2121-100ML
HONCE2121-100MLEA
266.86
EUR
HONCE2121-100ML
Multi-element solution 5 for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
10 mg/L each of B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr in HNO₃ 2%