VIS wavecal solution for ICP
Lieferant: Honeywell Chemicals
E2122-100ML
HONCE2122-100MLEA
190.54
EUR
HONCE2122-100ML
VIS wavecal solution for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
50 mg/L K, 10 mg/L each of La, Li, Mn, Na, Sr, 1 mg/L each of Ba, Ca in HNO₃ 2%