Primary Drinking Water Metals Certified Reference Material, 100 mg/L Ba, 10 mg/L each of Ag, As, Cr, Hg, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%, Fluka™
Lieferant: Honeywell Chemicals
E2164-100ML
HONCE2164-100MLEA
196.35
EUR
HONCE2164-100ML
Primary Drinking Water Metals Certified Reference Material, 100 mg/L Ba, 10 mg/L each of Ag, As, Cr, Hg, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%, Fluka™
Standards
Kationischer Standards
Mehrelement-Standards
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.